Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001278547.1 | NP_001265476.1 | ||||
NM_001278548.1 | NP_001265477.1 | |||||
NM_001323302.1 | NP_001310231.1 | |||||
NM_001323320.1 | NP_001310249.1 | |||||
NM_001323321.1 | NP_001310250.1 | |||||
NM_001323322.1 | NP_001310251.1 | |||||
NM_001323323.1 | NP_001310252.1 | |||||
NM_001323324.1 | NP_001310253.1 | |||||
NM_001323325.1 | NP_001310254.1 | |||||
NM_001323326.1 | NP_001310255.1 | |||||
NM_001323327.1 | NP_001310256.1 | |||||
NM_001323328.1 | NP_001310257.1 | |||||
NM_001323329.1 | NP_001310258.1 | |||||
NM_001323330.1 | NP_001310259.1 | |||||
NM_001323331.1 | NP_001310260.1 | |||||
NM_139046.3 | NP_620634.1 | |||||
NM_139049.3 | NP_620637.1 | |||||
NR_136583.1 | ||||||
NR_136584.1 | ||||||
NR_136585.1 | ||||||
GenBank mRNA | AA808177.1 | - | ||||
AB074280.1 | - | |||||
AB451231.1 | - | |||||
AB451271.1 | - | |||||
AB451354.1 | - | |||||
AB451398.1 | - | |||||
AK094000.1 | - | |||||
AK125150.1 | - | |||||
AK292523.1 | - | |||||
AL137667.1 | - | |||||
BC130570.1 | - | 40146601 | ||||
BC130572.1 | - | 40146603 | ||||
BC144063.1 | - | 9052583 | ||||
DA159335.1 | - | |||||
DQ234352.1 | - | |||||
ENST00000374182.0 | - | |||||
L26318.1 | - | |||||
U34822.1 | - | |||||
U35004.1 | - | |||||
U35005.1 | - |