Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3C2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_002646.3 | NP_002637.3 | ||||
XM_005245257.2 | XP_005245314.1 | |||||
XM_005245258.3 | XP_005245315.1 | |||||
XM_011509630.2 | XP_011507932.1 | |||||
XM_011509631.2 | XP_011507933.1 | |||||
XM_011509633.2 | XP_011507935.1 | |||||
XM_017001473.1 | XP_016856962.1 | |||||
XM_017001474.1 | XP_016856963.1 | |||||
XM_017001475.1 | XP_016856964.1 | |||||
GenBank mRNA | AB210006.1 | - | ||||
AK308153.1 | - | |||||
BC094741.1 | - | |||||
BC132876.1 | - | 40146898 | ||||
BC136645.1 | - | 9020635 | ||||
BC144342.1 | - | 9052862 | ||||
CR749201.1 | - | |||||
ENST00000367187.0 | - | |||||
F17716.1 | - | |||||
Y11312.1 | - | |||||
Y13892.1 | - |