Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angpt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_009641.1 | NP_033771.1 | ||||
GenBank mRNA | AF113707.1 | - | ||||
AK132988.1 | - | |||||
AK154379.1 | - | |||||
BC129965.1 | - | 40106226 | ||||
BC129966.1 | - | 40106227 |