Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_008366.3 | NP_032392.1 | ||||
GenBank mRNA | AF065914.1 | - | ||||
AF065915.1 | - | |||||
AF065916.1 | - | |||||
AF352786.1 | - | |||||
AF538059.1 | - | |||||
AF542383.1 | - | |||||
AF542384.1 | - | |||||
AF542385.1 | - | |||||
AY147902.1 | - | |||||
BC116845.1 | - | 40126164 | ||||
BC116873.1 | - | 40126192 | ||||
BX519775.1 | - | |||||
DQ836354.1 | - | |||||
K02292.1 | - | |||||
U41494.1 | - | |||||
U41504.1 | - | |||||
U41505.1 | - | |||||
U41506.1 | - | |||||
X01772.1 | - | |||||
X66058.1 | - | |||||
X73040.1 | - |