Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_008839.2 | NP_032865.2 | ||||
GenBank mRNA | AK018164.1 | - | ||||
AK029399.1 | - | |||||
AK039740.1 | - | |||||
AK051885.1 | - | |||||
AK053528.1 | - | |||||
AK078608.1 | - | |||||
AK151886.1 | - | |||||
AK164801.1 | - | |||||
BC089038.1 | - | 30692343 | ||||
BC130228.1 | - | 40141870 | ||||
U03279.1 | - |