Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dkk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_010051.3 | NP_034181.2 | ||||
GenBank mRNA | AF030433.1 | - | ||||
AK141693.1 | - | |||||
AK144119.1 | - | |||||
BC050189.1 | - | 30022799 | ||||
JN966751.1 | - |