Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Connexin 43 Antibodies

          Invitrogen

          Connexin 43 Monoclonal Antibody (3D8A5)

          Advanced Verification
          52 Published Figures
          53 References
          View all (42) Connexin 43 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Connexin 43 Monoclonal Antibody (3D8A5)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Published Figures (52)
          • Advanced Verification (1)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 55

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Connexin 43 Antibody (35-5000) in IHC

          Double immunofluorescent staining of mouse heart atrium using mouse anti-Connexin 43 monoclonal antibody (Product # 35-5000) (red) and Rabbit anti-Connexin 40 (C-term, Product # 36-4900) (green). Cx43 immunolocalizes to the intercalated discs throughout cardiac tissue, while Cx40 immunolabeling is localized to intercalated discs restricted to the conduction system. Image courtesy of James I. Nagy, PhD, Univ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Western Blot (WB)
          Connexin 43 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 43 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 43 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 43 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 43 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 43 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 43 Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry (IHC, IHC (P), ICC/IF)
          Connexin 43 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 43 Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
          Connexin 43 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 43 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 43 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 43 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 43 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 43 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Immunohistochemistry (IHC)
          Connexin 43 Antibody in Western Blot (WB)
          Connexin 43 Antibody in Western Blot (WB)
          Connexin 43 Antibody in Western Blot (WB)
          Connexin 43 Antibody in Western Blot (WB)
          Connexin 43 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Connexin 43 Antibody in Western Blot (WB)
          Connexin 43 Antibody in Western Blot (WB)
          Connexin 43 Antibody in In Situ Hybridization (ISH)
          Connexin 43 Antibody

          Product Details

          35-5000

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 16 publications 16 publications

          Immunohistochemistry (IHC)

          -
          View 18 publications 18 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          View 5 publications 5 publications

          Immunocytochemistry (ICC/IF)

          -
          View 8 publications 8 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          In Situ Hybridization (ISH)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          3D8A5

          Immunogen

          Synthetic peptide derived from rat Connexin 43.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533207

          Target Information

          Connexin 43 (Cx43) is a member of the gap junction protein family. Connexins assemble as a hexamer and are transported to the plasma membrane to create a hemichannel that can associate with hemichannels on nearby cells to create cell-to-cell channels. Clusters of these channels assemble to make gap junctions. Gap junction communication is important in development and regulation of cell growth. Phosphorylation of Cx43 is important in regulating assembly and function of gap junctions. Ser368 of Cx43 is phosphorylated by protein kinase C (PKC) after activation by phorbol esters, which decreases cell-to-cell communication. Src can interact with and phosphorylate Cx43 to alter gap junction communication. GFAP are membrane-spanning proteins that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Connexin 43 is the major protein of gap junctions in the heart, and gap junctions are thought to have a crucial role in the synchronized contraction of the heart and in embryonic development. Connexin 43 is also targeted by several protein kinases that regulate myocardial cell-cell coupling. A related intron-less connexin 43 pseudogene, GJA1P, has been mapped to chromosome 5. Mutations in the GFAP gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy, oculodentodigital dysplasia and heart malformations. Alternatively spliced transcript variants of GFAP have been found.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: alpha 1 connexin; connexin 43; Connexin-43; Cx-43; Cx43; CXA1; DFNB38; Gap junction 43 kDa heart protein; Gap junction alpha-1 protein; gap junction membrane channel protein alpha 1; ODD; ODOD; SDTY3; Syndactyly type III

          View more View less

          Gene Aliases: AU042049; AW546267; Cnx43; connexin43; Cx43; Cx43alpha1; Cxn-43; Gja-1; Gja1; Npm1

          View more View less

          UniProt ID: (Mouse) P23242, (Rat) P08050

          View more View less

          Entrez Gene ID: (Mouse) 14609, (Rat) 24392

          View more View less

          Function(s)
          signal transducer activity receptor binding gap junction channel activity protein binding ion transmembrane transporter activity SH3 domain binding protein domain specific binding transmembrane transporter activity PDZ domain binding beta-tubulin binding connexin binding scaffold protein binding gap junction channel activity involved in cell communication by electrical coupling gap junction
          Process(es)
          osteoblast differentiation in utero embryonic development neuron migration negative regulation of endothelial cell proliferation heart looping regulation of heart rate epithelial cell maturation lens development in camera-type eye chronic inflammatory response positive regulation of glomerular filtration atrial ventricular junction remodeling epicardial cell to mesenchymal cell transition apoptotic process cell communication signal transduction positive regulation of cytosolic calcium ion concentration cell-cell signaling heart development adult heart development regulation of heart contraction negative regulation of cell proliferation response to pH vascular transport positive regulation of gene expression negative regulation of gene expression cell communication by chemical coupling cell communication by electrical coupling positive regulation of cell communication by chemical coupling ATP transport regulation of bone mineralization positive regulation of insulin secretion ion transmembrane transport embryonic heart tube development embryonic digit morphogenesis positive regulation of I-kappaB kinase/NF-kappaB signaling skeletal muscle tissue regeneration cell-cell junction organization positive regulation of osteoblast differentiation positive regulation of protein catabolic process positive regulation of striated muscle tissue development positive regulation of vasoconstriction positive regulation of vasodilation regulation of bone remodeling blood vessel morphogenesis neuron projection morphogenesis protein oligomerization regulation of calcium ion transport transmembrane transport negative regulation of cardiac muscle cell proliferation milk ejection regulation of ventricular cardiac muscle cell membrane repolarization regulation of blood vessel remodeling regulation of atrial cardiac muscle cell membrane depolarization regulation of ventricular cardiac muscle cell membrane depolarization negative regulation of wound healing cardiac conduction establishment of epithelial cell polarity negative regulation of DNA biosynthetic process regulation of bicellular tight junction assembly positive regulation of behavioral fear response endothelium development response to glucose gap junction assembly response to fluid shear stress response to peptide hormone cellular response to mechanical stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline