Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Connexin 40 Antibodies

          Invitrogen

          Connexin 40 Polyclonal Antibody

          Advanced Verification
          28 Published Figures
          35 References
          View all (11) Connexin 40 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Connexin 40 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          • Published Figures (28)
          • Advanced Verification (3)
          Connexin 40 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Connexin 40 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 35

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Connexin 40 Antibody (36-4900) in IHC (P)

          Immunohistochemistry analysis of Connexin 40 showing staining in the membrane of paraffin-embedded human heart tissue (right) compared to a negative control without primary antibody (left). To expose target proteins, antigen retrieval was performed using 10mM sodium citrate (pH 6.0), microwaved for 8-15 min. Following antigen retrieval, tissues were blocked in 3% H2O2-methanol for 15 min at room temperature... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Connexin 40 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Connexin 40 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Connexin 40 Antibody in Western Blot (WB)
          Connexin 40 Antibody in Immunoprecipitation (IP)
          Connexin 40 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 40 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 40 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 40 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 40 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Connexin 40 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 40 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 40 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Connexin 40 Antibody in Immunohistochemistry (IHC)
          Connexin 40 Antibody in Immunohistochemistry (IHC)
          Connexin 40 Antibody in Immunohistochemistry (IHC)
          Connexin 40 Antibody in Immunohistochemistry (IHC)
          Connexin 40 Antibody in Immunohistochemistry (IHC)
          Connexin 40 Antibody in Immunohistochemistry (IHC)
          Connexin 40 Antibody in Western Blot (WB)
          Connexin 40 Antibody in Western Blot (WB)
          Connexin 40 Antibody in Western Blot (WB)
          Connexin 40 Antibody in Western Blot (WB)
          Connexin 40 Antibody in Western Blot (WB)
          Connexin 40 Antibody in Western Blot (WB)
          Connexin 40 Antibody in Western Blot (WB)
          Connexin 40 Antibody in Western Blot (WB)
          Connexin 40 Antibody in Western Blot (WB)
          Connexin 40 Antibody in Western Blot (WB)
          Connexin 40 Antibody in Western Blot (WB)
          Connexin 40 Antibody in Western Blot (WB)
          Connexin 40 Antibody in Western Blot (WB)
          Connexin 40 Antibody in Western Blot (WB)
          Connexin 40 Antibody
          Connexin 40 Antibody
          Connexin 40 Antibody
          Connexin 40 Polyclonal Antibody

          Product Details

          36-4900

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 14 publications 14 publications

          Immunohistochemistry (IHC)

          -
          View 11 publications 11 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:10-1:50
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:10-1:50
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 8 publications 8 publications

          Immunoprecipitation (IP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Dog, Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide derived from the C-terminal region of the mouse and rat Connexin 40 (Cx40) proteins. The human sequence at this location differs by only one amino acid.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533263

          Target Information

          Connexin 40 (Cx40), belongs to the connexin family of gap junction proteins, which are expressed in pancreas, kidney, skeletal muscle, liver, placenta and heart. Connexin 40 is highly expressed in vascular endothelial cells, and the mammalian heart (specifically in the atrial myocardium, and the His-Purkinje conduction system). At least three other connexins are expressed in the heart, including Cx37 (endocardium), Cx43 (atrial and ventricular myocardium, and distal parts of the conduction system) and Cx45 (throughout the heart). Cx40 and Cx43 frequently co-localize in cells of the cardiovascular system and participate in the formation of heterotypic gap junction channels.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: connexin 40; Connexin-40; Cx40; Gap junction alpha-5 protein; gap junction membrane channel protein alpha 5; gap junction protein alpha 5 40kDa

          View more View less

          Gene Aliases: 5730555N10Rik; ATFB11; Cnx40; CX40; Cxn-40; Gja-5; GJA5

          View more View less

          UniProt ID: (Human) P36382, (Rat) P28234, (Mouse) Q01231

          View more View less

          Entrez Gene ID: (Human) 2702, (Rat) 50563, (Mouse) 14613

          View more View less

          Function(s)
          gap junction channel activity protein binding gap junction hemi-channel activity connexin binding gap junction channel activity involved in SA node cell-atrial cardiac muscle cell electrical coupling gap junction channel activity involved in cardiac conduction electrical coupling gap junction channel activity involved in atrial cardiac muscle cell-AV node cell electrical coupling gap junction channel activity involved in AV node cell-bundle of His cell electrical coupling gap junction channel activity involved in bundle of His cell-Purkinje myocyte electrical coupling gap junction channel activity involved in Purkinje myocyte-ventricular cardiac muscle cell electrical coupling disordered domain specific binding gap junction
          Process(es)
          skeletal system development angiogenesis blood vessel development renal system process involved in regulation of systemic arterial blood pressure regulation of systemic arterial blood pressure negative regulation of glomerular filtration outflow tract morphogenesis endothelium development cardiac conduction system development mitral valve development pulmonary valve formation ventricular septum development atrial septum development septum primum development atrial ventricular junction remodeling potassium ion transport cell communication cell-cell signaling heart development cell communication by chemical coupling cell communication by electrical coupling regulation of cell communication by electrical coupling positive regulation of cell communication by chemical coupling gap junction assembly embryonic limb morphogenesis embryonic heart tube development foramen ovale closure vasodilation negative regulation of blood pressure positive regulation of vasoconstriction artery morphogenesis transmembrane transport regulation of cardiac muscle contraction regulation of ventricular cardiac muscle cell membrane repolarization regulation of atrial cardiac muscle cell membrane depolarization regulation of ventricular cardiac muscle cell membrane depolarization ventricular septum morphogenesis atrial septum morphogenesis cardiac conduction ventricular cardiac muscle cell action potential SA node cell action potential SA node cell to atrial cardiac muscle cell communication by electrical coupling atrial cardiac muscle cell to AV node cell communication by electrical coupling AV node cell to bundle of His cell communication by electrical coupling bundle of His cell to Purkinje myocyte communication by electrical coupling Purkinje myocyte to ventricular cardiac muscle cell communication by electrical coupling cell communication by electrical coupling involved in cardiac conduction cell communication involved in cardiac conduction AV node cell to bundle of His cell communication regulation of heart rate by cardiac conduction regulation of blood vessel diameter regulation of AV node cell action potential regulation of bundle of His cell action potential regulation of Purkinje myocyte action potential regulation of atrial cardiac muscle cell action potential regulation of renin secretion into blood stream regulation of membrane depolarization during cardiac muscle cell action potential vasomotion regulation of vasodilation positive regulation of vasodilation protein oligomerization atrial cardiac muscle cell action potential bundle of His cell to Purkinje myocyte communication
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-gkxk9:80/100.66.77.77:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline