Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 16
This gene encodes an adhesion protein that plays a role in the organization of adherens junctions and tight junctions in epithelial and endothelial cells. The protein is a calcium(2+)-independent cell-cell adhesion molecule that belongs to the immunoglobulin superfamily and has 3 extracellular immunoglobulin-like loops, a single transmembrane domain, and a cytoplasmic region. This protein acts as a receptor for glycoprotein D of herpes simplex viruses 1 and 2, and pseudorabies virus and mediates viral entry into epithelial and neuronal cells. Mutations in this gene cause cleft lip and palate/ectodermal dysplasia 1 syndrome as well as non-syndromic cleft lip with or without cleft palate. Alternative splicing results in multiple transcript variants encoding proteins with distinct C-termini.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD111; ectodermal dysplasia 4 (Margarita Island type); herpes simplex virus type 1 sensitivity; Herpes virus entry mediator C; Herpesvirus entry mediator C; Herpesvirus Ig-like receptor; HIgR; nectin 1; Nectin cell adhesion molecule 1; nectin); Nectin-1; nectin-1 alpha; nectin-1 delta; poliovirus receptor-like 1; poliovirus receptor-related 1 (herpesvirus entry mediator C; poliovirus receptor-related 1 (herpesvirus entry mediator C); poliovirus receptor-related 1 (herpesvirus entry mediator C; nectin); Poliovirus receptor-related protein 1
Gene Aliases: AI835281; AW549174; CD111; CLPED1; ED4; HIgR; HV1S; HVEC; nectin-1; NECTIN1; OFC7; PRR; PRR1; PVRL1; PVRR; PVRR1; SK-12
UniProt ID: (Human) Q15223, (Mouse) Q9JKF6
Entrez Gene ID: (Human) 5818, (Rat) 192183, (Mouse) 58235
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support