Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: LEIGRFDPER GRGAAPCQAV EIPMCRGIGY NLTRMPNLLG HTSQGEAAAE LAEFAPLVQY GCHSHLRFFL CSLYAPMCTD QVSTPIPACR PMCEQARLRC APIMEQFNFG WPDSLDCARL PTRNDPHALC MEAPENATAG PAEPHKGLGM LPVAPRPARP PGDLGPGAGG SGTCENREKF QYVEKSRSCA PRCGPGVEVF WSRRDKDFAL VWMAVWSALC FFSTA
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD9 gene is located within the Williams syndrome common deletion region of chromosome 7, and heterozygous deletion of the FZD9 gene may contribute to the Williams syndrome phenotype. FZD9 is expressed predominantly in brain, testis, eye, skeletal muscle, and kidney.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD349; CD349 antigen; Frizzled 3; frizzled 9, seven transmembrane spanning receptor; frizzled family receptor 9; frizzled homolog 9; Frizzled-3; Frizzled-9; Frizzled3; Frizzled9; Fz-9; FzE6; hFz9
Gene Aliases: CD349; FZD3; FZD9
UniProt ID: (Human) O00144
Entrez Gene ID: (Human) 8326
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support