Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
LOC100507346
Transcript:
ENST00000331920.0
Assay Name:
Mutation genome location:
chr.9 95467171-95467343 on build GRCh38
Gene:
PTCH1
Transcript:
ENST00000331920.0
Assay Name:
Mutation genome location:
chr.9 95467171-95467343 on build GRCh38
Assay ID
Hs00523355_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Less than 200 bp
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
branching involved in ureteric bud morphogenesis
in utero embryonic development
cell fate determination
neural tube closure
heart morphogenesis
smoothened signaling pathway
regulation of mitotic cell cycle
brain development
regulation of smoothened signaling pathway
response to mechanical stimulus
organ morphogenesis
dorsal/ventral pattern formation
epidermal cell fate specification
response to chlorate
positive regulation of cholesterol efflux
protein processing
spinal cord motor neuron differentiation
neural tube patterning
neural plate axis specification
embryonic limb morphogenesis
response to estradiol
response to retinoic acid
regulation of protein localization
limb morphogenesis
hindlimb morphogenesis
negative regulation of multicellular organism growth
response to drug
glucose homeostasis
negative regulation of sequence-specific DNA binding transcription factor activity
keratinocyte proliferation
positive regulation of epidermal cell differentiation
negative regulation of osteoblast differentiation
negative regulation of smoothened signaling pathway
positive regulation of transcription, DNA-templated
embryonic organ development
negative regulation of epithelial cell proliferation
negative regulation of cell division
pharyngeal system development
mammary gland duct morphogenesis
mammary gland epithelial cell differentiation
smoothened signaling pathway involved in dorsal/ventral neural tube patterning
cell differentiation involved in kidney development
somite development
cellular response to cholesterol
renal system development
cell proliferation involved in metanephros development
protein targeting to plasma membrane
patched binding
smoothened binding
protein binding
hedgehog receptor activity
heparin binding
cholesterol binding
cyclin binding
protein complex binding
hedgehog family protein binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-fcf5f9d6d-svqt6:80/100.66.77.77:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0