Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PAK3 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001128166.2 | NP_001121638.1 | - | - | - |
NM_001128167.2 | NP_001121639.1 | - | - | - | |
NM_001128168.2 | NP_001121640.1 | - | - | - | |
NM_001128172.2 | NP_001121644.1 | - | - | - | |
NM_001128173.2 | NP_001121645.1 | - | - | - | |
NM_001324325.1 | NP_001311254.1 | - | - | - | |
NM_001324326.1 | NP_001311255.1 | - | - | - | |
NM_001324327.1 | NP_001311256.1 | - | - | - | |
NM_001324328.1 | NP_001311257.1 | - | - | - | |
NM_001324329.1 | NP_001311258.1 | - | - | - | |
NM_001324330.1 | NP_001311259.1 | - | - | - | |
NM_001324331.1 | NP_001311260.1 | - | - | - | |
NM_001324332.1 | NP_001311261.1 | - | - | - | |
NM_001324333.1 | NP_001311262.1 | - | - | - | |
NM_001324334.1 | NP_001311263.1 | - | - | - | |
NM_002578.4 | NP_002569.1 | - | - | - | |
NR_136747.1 | - | - | - | ||
NR_136748.1 | - | - | - | ||
XM_005262132.2 | XP_005262189.1 | - | - | - | |
XM_005262133.3 | XP_005262190.1 | - | - | - | |
XM_006724654.2 | XP_006724717.1 | - | - | - | |
XM_006724655.2 | XP_006724718.1 | - | - | - | |
XM_011530962.1 | XP_011529264.1 | - | - | - | |
XM_011530967.2 | XP_011529269.1 | - | - | - | |
XM_011530968.2 | XP_011529270.1 | - | - | - | |
XM_011530969.1 | XP_011529271.1 | - | - | - | |
XM_011530971.2 | XP_011529273.1 | - | - | - | |
XM_017029557.1 | XP_016885046.1 | - | - | - | |
XM_017029558.1 | XP_016885047.1 | - | - | - | |
XM_017029559.1 | XP_016885048.1 | - | - | - | |
XM_017029560.1 | XP_016885049.1 | - | - | - | |
XM_017029561.1 | XP_016885050.1 | - | - | - | |
XM_017029562.1 | XP_016885051.1 | - | - | - | |
XM_017029563.1 | XP_016885052.1 | - | - | - | |
GenBank | AB102659.1 | BAC81128.1 | - | - | - |
AF068864.1 | AAC36097.1 | - | - | - | |
AF155651.1 | AAF67008.1 | - | - | - | |
AK128022.1 | - | - | - | ||
AK290504.1 | - | - | - | ||
AK315284.1 | - | - | - | ||
AM943850.1 | - | - | - | ||
AM943851.1 | - | - | - | ||
AM943852.1 | - | - | - | ||
BC117353.1 | - | - | - | ||
BC117355.1 | - | - | - | ||
BC143544.1 | - | - | - | ||
EF491800.1 | - | - | - | ||
ENST00000446737.0 | - | - | - |
Literature Links: |
PAK3 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |