Display Info New header:
tfpt-header-footer.tfcom-eks-na-comerce-gateway-prd.commerceprod.thermofisher.net cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@f993913
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:/anonymous enableSimpleHeaderValue:
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Copy Number Variation
Search All
Popular Product Areas
TaqMan Assays
Gene Expression
Microbe & ABR Detection
Copy Number Variation
Mutation Detection
miRNA
SNP Genotyping
Primary Antibodies
Secondary Antibodies
Isotype Controls
Proteins & Peptides
ELISA Kits
All Documents & Support
Certificates
SDS
Manuals & Protocols
Product FAQs
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs00768454_cn
See other AP3S2 CNV Assays ›
Gene Symbol
AP3S2
Assay Reference Genome
Location
Chr.15:89834095 on build GRCh38
Cytoband
15q26.1
Assay ID
Hs00768454_cn
Size
S: 360 rxns
M: 750 rxns
L: 2,900 rxns
Availability
Made To Order
Catalog #
4400291
Price (USD)
457.00
Your Price
Online offer (USD):
424.65
424.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
Displaying...
Build 38.2 Human Chr.15: Hs00768454_cn
×
Created with Raphaël 2.1.2
ANPEP
AP3S2
C15orf38
-AP3S2
ARPIN
ANPEP
AP3S2
C15orf38
-AP3S2
MIR5094
MIR5009
ARPIN
89,580,0
00
89,590,0
00
89,600,0
00
89,610,0
00
89,620,0
00
89,630,0
00
89,640,0
00
89,650,0
00
89,660,0
00
89,670,0
00
89,680,0
00
89,690,0
00
89,700,0
00
89,710,0
00
89,720,0
00
89,730,0
00
89,740,0
00
89,750,0
00
89,760,0
00
89,770,0
00
89,780,0
00
89,790,0
00
89,800,0
00
89,810,0
00
89,820,0
00
89,830,0
00
89,840,0
00
89,850,0
00
89,860,0
00
89,870,0
00
89,880,0
00
89,890,0
00
89,900,0
00
89,910,0
00
89,920,0
00
89,930,0
00
89,940,0
00
89,950,0
00
89,960,0
00
89,970,0
00
89,980,0
00
89,990,0
00
90,000,0
00
90,010,0
00
90,020,0
00
90,030,0
00
90,040,0
00
90,050,0
00
90,060,0
00
90,070,0
00
90,080,0
00
90,090,0
00
90,100,0
00
90,110,0
00
90,120,0
00
90,130,0
00
90,140,0
00
90,150,0
00
90,160,0
00
90,170,0
00
Show DGVs
Back To Top
Assay Details
Target Gene Details
Entrez Gene ID:
10239
Gene Name:
adaptor related protein complex 3 sigma 2 subunit
Gene Aliases:
AP3S3, sigma3b
Location:
Chr.15:89830599-89894385 on Build GRCh38
Assay Gene Location:
Within Exon 7
Gene Symbol
Transcript Accession
Exon Location
Assay Transcript Location
Protein ID
AP3S2
NM_005829.4
6
2438
NP_005820.1
NR_023361.1
7
2557
NR_037582.1
6
2434
BC002785.2
6
2023
AAH02785.1
BC007773.2
AAH07773.1
DA337392.1
DR158459.1
1
233
Target Gene Details
Entrez Gene ID:
100526783
Gene Name:
C15orf38-AP3S2 readthrough
Gene Aliases:
-
Location:
Chr.15:89830599-89912990 on Build GRCh38
Assay Gene Location:
Within Exon 10
Gene Symbol
Transcript Accession
Exon Location
Assay Transcript Location
Protein ID
C15orf38-AP3S2
NM_001199058.1
10
2760
NP_001185987.1
Target Copy Number Variation Details
DGV Version:
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location
CNV
Subtype
Genes
nsv1661
Chr.15:89804782 - 89839712 on Build GRCh38
Insertion
AP3S2
ANPEP
C15orf38-AP3S2
Back To Top
More Information
Set Membership:
Intragenic
Exonic
DGV Variation
Panther Classification:
Molecular Function -
vesicle coat protein
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
anterograde axonal transport
anterograde synaptic vesicle transport
protein transporter activity
Back To Top
Related Products
TaqMan
®
Genotyping Master Mix
TaqPath
®
ProAmp
®
Master Mix
TaqMan
®
Universal Master Mix II, no UNG
TaqMan
®
Gene Expression Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Universal PCR Master Mix, No AmpErase UNG
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:/anonymous
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-59d6677d6-wpz24:80/100.66.78.181:80.
git-commit: 1e3318a2800c8d0a8829a508293490082e5b9833
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.30.0-Offline