Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 4609 |
Gene Name: | v-myc avian myelocytomatosis viral oncogene homolog |
Gene Aliases: |
MRTL, MYCC, bHLHe39, c-Myc |
Location: |
Chr.8:127736069-127741434 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYC | NM_002467.4 | 3 | 1488 | NP_002458.2 |
AK303921.1 | 4 | 1270 | ||
AK312883.1 | 3 | 1321 | ||
BC000141.1 | 3 | 1299 | ||
BC000917.2 | 3 | 1315 | ||
BC058901.1 | 3 | 1001 | ||
BT019768.1 | 2 | 918 | AAV38573.1 | |
K02276.1 | 3 | 1338 | AAA36340.1 | |
KC782559.1 | 2 | 191 | ||
V00568.1 | 3 | 1476 | CAA23831.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv516713 | Chr.8:127727525 - 127755908 on Build GRCh38 | Gain+Loss |
![]() ![]() |
nsv831458 | Chr.8:127702603 - 127875740 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv2752208 | Chr.8:127632573 - 127895715 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
esv3618781 | Chr.8:127738039 - 127746724 on Build GRCh38 | Gain |
![]() |
Set Membership: |
![]() ![]() ![]() |