Display Info New header:
tfpt-header-footer.tfcom-eks-na-comerce-gateway-prd.commerceprod.thermofisher.net cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@53eddcab
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:/anonymous enableSimpleHeaderValue:
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search All
Popular Product Areas
TaqMan Assays
Gene Expression
Microbe & ABR Detection
Copy Number Variation
Mutation Detection
miRNA
SNP Genotyping
Primary Antibodies
Secondary Antibodies
Isotype Controls
Proteins & Peptides
ELISA Kits
All Documents & Support
Certificates
SDS
Manuals & Protocols
Product FAQs
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs00297871_cn
See other NR1H2 CNV Assays ›
Gene Symbol
NR1H2
Assay Reference Genome
Location
Chr.19:50379161 on build GRCh38
Cytoband
19q13.33
Assay ID
Hs00297871_cn
Size
S: 360 rxns
M: 750 rxns
L: 2,900 rxns
Availability
Made To Order
Catalog #
4400291
Price (USD)
457.00
Your Price
Online offer (USD):
424.65
424.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
Displaying...
Build 38.2 Human Chr.19: Hs00297871_cn
×
Created with Raphaël 2.1.2
NR1H2
POLD1
NR1H2
POLD1
50,356,0
00
50,357,0
00
50,358,0
00
50,359,0
00
50,360,0
00
50,361,0
00
50,362,0
00
50,363,0
00
50,364,0
00
50,365,0
00
50,366,0
00
50,367,0
00
50,368,0
00
50,369,0
00
50,370,0
00
50,371,0
00
50,372,0
00
50,373,0
00
50,374,0
00
50,375,0
00
50,376,0
00
50,377,0
00
50,378,0
00
50,379,0
00
50,380,0
00
50,381,0
00
50,382,0
00
50,383,0
00
50,384,0
00
50,385,0
00
50,386,0
00
50,387,0
00
50,388,0
00
50,389,0
00
50,390,0
00
50,391,0
00
50,392,0
00
50,393,0
00
50,394,0
00
50,395,0
00
50,396,0
00
50,397,0
00
50,398,0
00
50,399,0
00
50,400,0
00
50,401,0
00
50,402,0
00
50,403,0
00
Show DGVs
Back To Top
Assay Details
Target Gene Details
Entrez Gene ID:
7376
Gene Name:
nuclear receptor subfamily 1 group H member 2
Gene Aliases:
LXR-b, LXRB, NER, NER-I, RIP15, UNR
Location:
Chr.19:50376423-50383028 on Build GRCh38
Assay Gene Location:
Overlaps Exon 7 - Intron 7
Gene Symbol
Transcript Accession
Exon Location
Assay Transcript Location
Protein ID
NR1H2
NM_001256647.1
NP_001243576.1
NM_007121.5
NP_009052.3
AB307697.1
AK091535.1
AK290855.1
AK297978.1
BC007790.1
AAH07790.1
BC033500.1
AAH33500.1
BC047750.2
AAH47750.1
BC074500.1
AAH74500.1
CR456706.1
CAG32987.1
CR749648.1
CAH18442.1
HQ692835.1
U07132.1
AAA61783.1
U14534.1
AAA58594.1
Target Copy Number Variation Details
DGV Version:
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location
CNV
Subtype
Genes
nsv1066389
Chr.19:50319151 - 50453251 on Build GRCh38
Gain
NR1H2
KCNC3
POLD1
MYBPC2
NAPSA
NAPSB
SPIB
Back To Top
More Information
Set Membership:
Intragenic
Non-exonic
DGV Variation
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
negative regulation of macrophage derived foam cell differentiation
positive regulation of triglyceride biosynthetic process
positive regulation of cholesterol efflux
positive regulation of lipid storage
negative regulation of cholesterol storage
positive regulation of cellular protein metabolic process
negative regulation of lipid transport
positive regulation of cholesterol transport
cholesterol homeostasis
steroid hormone mediated signaling pathway
cellular lipid metabolic process
positive regulation of fatty acid biosynthetic process
negative regulation of proteolysis
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
retinoic acid receptor signaling pathway
negative regulation of pinocytosis
positive regulation of lipoprotein lipase activity
negative regulation of interferon-gamma-mediated signaling pathway
positive regulation of high-density lipoprotein particle assembly
positive regulation of pancreatic juice secretion
positive regulation of secretion of lysosomal enzymes
regulation of cholesterol homeostasis
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor activity, sequence-specific transcription regulatory region DNA binding
DNA binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
protein binding
zinc ion binding
apolipoprotein A-I receptor binding
retinoid X receptor binding
ATPase binding
Back To Top
Related Products
TaqMan
®
Genotyping Master Mix
TaqPath
®
ProAmp
®
Master Mix
TaqMan
®
Universal Master Mix II, no UNG
TaqMan
®
Gene Expression Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Universal PCR Master Mix, No AmpErase UNG
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:/anonymous
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-59d6677d6-wpz24:80/100.66.78.181:80.
git-commit: 1e3318a2800c8d0a8829a508293490082e5b9833
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.30.0-Offline