Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
  • Special Offers
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Microbe & ABR Detection
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            • Home
            • › Search Tool
            • › Search Results
            • › Hs01255090_cn
            See other ROM1 CNV Assays ›
            Gene Symbol
            ROM1
            Assay Reference Genome
            Location

            Chr.11:62615082 on build GRCh38
            Cytoband
            11q12.3
            Assay ID Hs01255090_cn
            Size
            Availability Made To Order
            Catalog # 4400291
            Price (USD) 457.00
            Your Price
            Online offer (USD):424.65
            424.65
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data.
            ×
            Back To Top

            Assay Details

            Target Gene Details

            Entrez Gene ID:

            6094

            Gene Name:

            retinal outer segment membrane protein 1

            Gene Aliases:

            ROM, ROSP1, RP7, TSPAN23

            Location:

            Chr.11:62612741-62615120 on Build GRCh38

            Assay Gene Location:

            Overlaps - Exon 3
            Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
            ROM1 NM_000327.3 NP_000318.1
            BC008100.1 AAH08100.1
            BU731899.1
            L07894.1 AAA60274.1

            Target Copy Number Variation Details

            DGV Version:

            Release date: 2016-05-15, GRCh GRCh38
            Target
            Variation
            Location CNV
            Subtype
            Genes
            esv3584599 Chr.11:62495270 - 62630578 on Build GRCh38 Gain EEF1G MIR6747 EML3 ROM1 AHNAK B3GAT3 MTA2 TUT1 GANAB
            nsv825939 Chr.11:62558611 - 62643105 on Build GRCh38 Gain EEF1G MIR6747 EML3 ROM1 B3GAT3 MTA2 TUT1 GANAB
            nsv832182 Chr.11:62528103 - 62695376 on Build GRCh38 Gain+Loss ROM1 UQCC3 B3GAT3 HNRNPUL2-BSCL2 LRRN4CL C11orf98 MTA2 EEF1G MIR6747 EML3 SNORA57 BSCL2 AHNAK METTL12 INTS5 UBXN1 LBHD1 TUT1 GANAB
            nsv555174 Chr.11:62566624 - 62729047 on Build GRCh38 Gain ROM1 HNRNPUL2 UQCC3 B3GAT3 HNRNPUL2-BSCL2 LRRN4CL C11orf98 MTA2 TTC9C EEF1G MIR6747 EML3 SNORA57 BSCL2 METTL12 INTS5 UBXN1 GNG3 LBHD1 TUT1 GANAB

            Back To Top

            More Information


            Set Membership:

            Intragenic Non-exonic DGV Variation

            Gene Ontology Categories:

            Function(s)

            cell adhesion
            visual perception
            regulation of gene expression
            camera-type eye photoreceptor cell differentiation
            retina vasculature development in camera-type eye

            Back To Top

            Related Products

            • TaqMan® Genotyping Master Mix
            • TaqPath®ProAmp® Master Mix
            • TaqMan® Universal Master Mix II, no UNG
            • TaqMan® Gene Expression Master Mix
            • TaqMan® Universal PCR Master Mix
            • TaqMan® Universal PCR Master Mix, No AmpErase UNG
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-6ff95d844f-xpgds:80/100.66.76.20:80.
            git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.28.0-Offline