Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Microbe & ABR Detection
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            • Home
            • › Search Tool
            • › Search Results
            • › Hs02656958_cn
            See other CNP CNV Assays ›
            Gene Symbol
            CNP
            Assay Reference Genome
            Location

            Chr.17:41975703 on build GRCh38
            Cytoband
            17q21.2
            Assay ID Hs02656958_cn
            Size
            Availability Made To Order
            Catalog # 4400291
            Price
            Your Price
            Online offer:
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data..
            ×
            Back To Top

            Assay Details

            Target Gene Details

            Entrez Gene ID:

            1267

            Gene Name:

            2',3'-cyclic nucleotide 3' phosphodiesterase

            Gene Aliases:

            CNP1

            Location:

            Chr.17:41966741-41977736 on Build GRCh38

            Assay Gene Location:

            Within Exon 5
            Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
            CNP NM_033133.4 4 3189 NP_149124.3
            AK098048.1
            AK124861.1 1 539

            Target Copy Number Variation Details

            DGV Version:

            Release date: 2016-05-15, GRCh GRCh38
            Target
            Variation
            Location CNV
            Subtype
            Genes
            nsv953897 Chr.17:41966883 - 41977382 on Build GRCh38 Deletion CNP DNAJC7
            nsv509658 Chr.17:41927491 - 42002888 on Build GRCh38 Insertion ACLY TTC25 CNP DNAJC7

            Back To Top

            More Information


            Set Membership:

            Intragenic Exonic DGV Variation

            Panther Classification:

            Molecular Function -

            hydrolase phosphodiesterase metabolite interconversion enzyme

            Gene Ontology Categories:

            Function(s) Process(es)

            microtubule cytoskeleton organization
            chemical synaptic transmission
            axonogenesis
            aging
            adult locomotory behavior
            cyclic nucleotide catabolic process
            response to toxic substance
            substantia nigra development
            forebrain development
            response to lipopolysaccharide
            regulation of mitochondrial membrane permeability
            oligodendrocyte differentiation
            RNA binding
            2',3'-cyclic-nucleotide 3'-phosphodiesterase activity
            cyclic nucleotide binding

            Back To Top

            Related Products

            • TaqMan® Genotyping Master Mix
            • TaqPath®ProAmp® Master Mix
            • TaqMan® Universal Master Mix II, no UNG
            • TaqMan® Gene Expression Master Mix
            • TaqMan® Universal PCR Master Mix
            • TaqMan® Universal PCR Master Mix, No AmpErase UNG
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-bqh2p:80/100.66.77.138:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0