Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 215 |
Gene Name: | ATP binding cassette subfamily D member 1 |
Gene Aliases: |
ABC42, ALD, ALDP, AMN |
Location: |
Chr.X:153724851-153744762 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABCD1 | NM_000033.3 | NP_000024.2 | ||
AY421736.1 | AAR07956.1 | |||
AY421737.1 | AAR07957.1 | |||
AY421738.1 | AAR07958.1 | |||
BC015541.1 | AAH15541.1 | |||
BC025358.1 | AAH25358.1 | |||
Z21876.1 | CAA79922.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv21656 | Chr.X:153687433 - 153750282 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() |
Set Membership: |
![]() ![]() ![]() ![]() |