Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2746 |
Gene Name: | glutamate dehydrogenase 1 |
Gene Aliases: |
GDH, GDH1, GLUD |
Location: |
Chr.10:87050202-87095047 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLUD1 | NM_001318900.1 | NP_001305829.1 | ||
NM_001318901.1 | NP_001305830.1 | |||
NM_001318902.1 | NP_001305831.1 | |||
NM_001318904.1 | NP_001305833.1 | |||
NM_001318905.1 | NP_001305834.1 | |||
NM_001318906.1 | NP_001305835.1 | |||
NM_005271.4 | NP_005262.1 | |||
AB209406.1 | BAD92643.1 | |||
AK094782.1 | ||||
AK122685.1 | ||||
AK222818.1 | BAD96538.1 | |||
AK294685.1 | ||||
AK297441.1 | ||||
AK297456.1 | ||||
AK303458.1 | ||||
AK311722.1 | ||||
BC040132.1 | AAH40132.1 | |||
BC112946.1 | AAI12947.1 | |||
GU727646.1 | ||||
J03248.1 | AAA52523.1 | |||
M20867.1 | AAA52526.1 | |||
M37154.1 | AAA52525.1 | |||
X07674.1 | CAA30521.1 | |||
X07769.1 | CAA30598.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428236 | Chr.10:86866859 - 87550005 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2759774 | Chr.10:86755301 - 87550005 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv831939 | Chr.10:86984277 - 87154254 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
Set Membership: |
![]() ![]() ![]() ![]() |