Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 57460 |
Gene Name: | protein phosphatase, Mg2+/Mn2+ dependent 1H |
Gene Aliases: |
ARHCL1, NERPP-2C, URCC2 |
Location: |
Chr.12:62643982-62935135 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPM1H | NM_020700.1 | NP_065751.1 | ||
XM_011538578.2 | XP_011536880.1 | |||
XM_011538579.1 | XP_011536881.1 | |||
XM_017019676.1 | XP_016875165.1 | |||
AB032983.1 | BAA86471.1 | |||
AB084258.1 | ||||
AK023315.1 | ||||
BC157843.1 |
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |