Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Microbe & ABR Detection
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            • Home
            • › Search Tool
            • › Search Results
            • › Hs06838302_cn
            See other MTAP CNV Assays ›
            Gene Symbol
            MTAP
            Assay Reference Genome
            Location

            Chr.9:21805102 on build GRCh38
            Cytoband
            9p21.3
            Assay ID Hs06838302_cn
            Size
            Availability Made To Order
            Catalog # 4400291
            Price
            Your Price
            Online offer:
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data.
            ×
            Back To Top

            Assay Details

            Target Gene Details

            Entrez Gene ID:

            4507

            Gene Name:

            methylthioadenosine phosphorylase

            Gene Aliases:

            BDMF, DMSFH, DMSMFH, HEL-249, LGMBF, MSAP, c86fus

            Location:

            Chr.9:21802636-21865971 on Build GRCh38

            Assay Gene Location:

            Within Intron 1
            Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
            MTAP NM_002451.3 NP_002442.2
            AF109294.1 AAD19641.1
            AK300592.1
            AK309365.1
            AY712791.1 AAU04442.1
            BC012316.1
            BC026106.1 AAH26106.1
            CR541670.1 CAG46471.1
            CR541710.1 CAG46511.1
            FJ224338.1
            HE654772.1
            HE654773.1
            HE654774.1
            HE654775.1
            HE654776.1
            HE654777.1
            L40432.1 AAG38871.1
            U22233.1 AAA81646.1

            Target Copy Number Variation Details

            DGV Version:

            Release date: 2016-05-15, GRCh GRCh38
            Target
            Variation
            Location CNV
            Subtype
            Genes
            nsv1026981 Chr.9:21748203 - 21811698 on Build GRCh38 Gain MTAP
            esv2764150 Chr.9:20659506 - 22349184 on Build GRCh38 Gain+Loss IFNA7 IFNW1 IFNA8 IFNA4 IFNA2 CDKN2B-AS1 KLHL9 IFNE IFNA10 IFNA6 FOCAD MIR491 IFNA13 CDKN2B IFNA1 MIR31 IFNA16 MTAP IFNA17 IFNB1 ERVFRD-3 MIR31HG IFNA5 CDKN2A-AS1 IFNA21 CDKN2A IFNA22P HACD4 FOCAD-AS1 IFNA14
            nsv8426 Chr.9:21763811 - 22176201 on Build GRCh38 Loss MTAP ERVFRD-3 CDKN2B-AS1 CDKN2A-AS1 CDKN2A CDKN2B

            Back To Top

            More Information


            Set Membership:

            Intragenic Intronic Non-exonic DGV Variation

            Panther Classification:

            Molecular Function -

            nucleotide kinase

            Gene Ontology Categories:

            Function(s) Process(es)

            nucleobase-containing compound metabolic process
            purine ribonucleoside salvage
            nicotinamide riboside catabolic process
            L-methionine biosynthetic process from methylthioadenosine
            methylation
            response to testosterone
            purine-containing compound salvage
            phosphorylase activity
            protein binding
            S-methyl-5-thioadenosine phosphorylase activity

            Back To Top

            Related Products

            • TaqMan® Genotyping Master Mix
            • TaqPath®ProAmp® Master Mix
            • TaqMan® Universal Master Mix II, no UNG
            • TaqMan® Gene Expression Master Mix
            • TaqMan® Universal PCR Master Mix
            • TaqMan® Universal PCR Master Mix, No AmpErase UNG
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mfpfg:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0