Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTLA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001037631.2 | NP_001032720.1 | ||||
NM_005214.4 | NP_005205.2 | |||||
GenBank mRNA | AF414120.1 | - | ||||
AF486806.1 | - | |||||
AK313732.1 | - | |||||
AY209009.1 | - | |||||
AY792514.1 | - | |||||
AY999702.1 | - | |||||
BC069566.1 | - | 7262243 | ||||
BC070162.1 | - | 30417685 | ||||
BC074842.2 | - | 30915552 | ||||
BC074893.2 | - | 30915247 | ||||
BG058720.1 | - | |||||
CD639535.1 | - | |||||
DQ785106.1 | - | |||||
ENST00000302823.0 | - | |||||
L15006.1 | - | |||||
U90273.1 | - |