Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPP1CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_002709.2 | NP_002700.1 | ||||
NM_206876.1 | NP_996759.1 | |||||
GenBank mRNA | AF092905.1 | - | ||||
AI147919.1 | - | |||||
AK295957.1 | - | |||||
AK303682.1 | - | |||||
AK312329.1 | - | |||||
AK316532.1 | - | |||||
BC002697.2 | - | 3609386 | ||||
BC012045.1 | - | 3631338 | ||||
BE047402.1 | - | |||||
BI551898.1 | - | |||||
BM835970.1 | - | |||||
BQ933435.1 | - | |||||
BT019744.1 | - | |||||
BU740022.1 | - | |||||
BX477003.1 | - | |||||
BX647295.1 | - | |||||
BX647970.1 | - | |||||
CB111140.1 | - | |||||
CD656807.1 | - | |||||
CD722576.1 | - | |||||
CR542263.1 | - | |||||
CR542285.1 | - | |||||
ENST00000358506.0 | - | |||||
EU668339.1 | - | |||||
X80910.1 | - |