Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000660.5 | NP_000651.3 | ||||
XM_011527242.1 | XP_011525544.1 | |||||
GenBank mRNA | AI304490.1 | - | ||||
AK291907.1 | - | |||||
AK307742.1 | - | |||||
AY820829.1 | - | |||||
BC000125.1 | - | 3351664 | ||||
BC001180.1 | - | 3356605 | ||||
BC022242.1 | - | 4399762 | ||||
BG115704.1 | - | |||||
BP267938.1 | - | |||||
BT007245.1 | - | |||||
DA935180.1 | - | |||||
ENST00000221930.0 | - | |||||
M38449.1 | - | |||||
X02812.1 | - |