Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100282701 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001155608.1 | NP_001149080.1 | ||||
GenBank mRNA | BT033731.1 | - | ||||
BT040831.1 | - | |||||
BT068341.1 | - | |||||
EU960247.1 | - | |||||
EU961986.1 | - |