Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_005417.4 | NP_005408.1 | ||||
NM_198291.2 | NP_938033.1 | |||||
XM_011529013.2 | XP_011527315.1 | |||||
XM_017028024.1 | XP_016883513.1 | |||||
XM_017028025.1 | XP_016883514.1 | |||||
XM_017028026.1 | XP_016883515.1 | |||||
XM_017028027.1 | XP_016883516.1 | |||||
GenBank mRNA | AF272982.1 | - | ||||
AK024281.1 | - | |||||
AK091756.1 | - | |||||
AW135290.1 | - | |||||
BC011566.1 | - | 4548399 | ||||
BC051270.1 | - | 4871614 | ||||
CD364525.1 | - | |||||
ENST00000445403.0 | - | |||||
HY016645.1 | - | |||||
Z18364.1 | - |