Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001256090.1 | NP_001243019.1 | ||||
NM_001256091.1 | NP_001243020.1 | |||||
NM_001256092.1 | NP_001243021.1 | |||||
NM_001256093.1 | NP_001243022.1 | |||||
NM_001256094.1 | NP_001243023.1 | |||||
NM_001880.3 | NP_001871.2 | |||||
NR_045768.1 | ||||||
NR_045769.1 | ||||||
NR_045770.1 | ||||||
NR_045771.1 | ||||||
NR_045772.1 | ||||||
NR_045773.1 | ||||||
NR_045774.1 | ||||||
GenBank mRNA | AF283776.2 | - | ||||
AI200584.1 | - | |||||
AK128731.1 | - | |||||
AL050192.1 | - | |||||
AY029364.1 | - | |||||
BC026175.1 | - | |||||
BC107698.1 | - | 6456836 | ||||
BC130335.1 | - | 40146366 | ||||
BC130337.1 | - | 40146368 | ||||
BX648469.1 | - | |||||
DA995464.1 | - | |||||
DQ003036.1 | - | |||||
DQ003037.1 | - | |||||
DQ003038.1 | - | |||||
DQ003039.1 | - | |||||
DQ003040.1 | - | |||||
DQ003041.1 | - | |||||
DQ003042.1 | - | |||||
DQ003043.1 | - | |||||
DQ003044.1 | - | |||||
DQ003045.1 | - | |||||
DQ003046.1 | - | |||||
DQ003047.1 | - | |||||
DQ003048.1 | - | |||||
DQ003049.1 | - | |||||
ENST00000264110.0 | - | |||||
M31630.1 | - | |||||
X15875.1 | - |