Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IL4R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000418.3 | NP_000409.1 | ||||
NM_001257406.1 | NP_001244335.1 | |||||
NM_001257407.1 | NP_001244336.1 | |||||
NM_001257997.1 | NP_001244926.1 | |||||
XM_005255308.2 | XP_005255365.1 | |||||
XM_006721043.1 | XP_006721106.1 | |||||
XM_011545825.1 | XP_011544127.1 | |||||
XM_011545826.2 | XP_011544128.1 | |||||
XM_011545827.2 | XP_011544129.1 | |||||
XM_011545828.2 | XP_011544130.1 | |||||
XM_011545829.1 | XP_011544131.1 | |||||
XM_011545830.2 | XP_011544132.1 | |||||
XM_011545831.1 | XP_011544133.1 | |||||
XM_011545832.1 | XP_011544134.1 | |||||
XM_011545833.2 | XP_011544135.1 | |||||
XM_011545834.2 | XP_011544136.1 | |||||
XM_017023211.1 | XP_016878700.1 | |||||
GenBank mRNA | AA838318.1 | - | ||||
AB102798.1 | - | |||||
AK223591.1 | - | |||||
AK303255.1 | - | |||||
AK314863.1 | - | |||||
BC110343.1 | - | |||||
BC136365.1 | - | 9020352 | ||||
BC151131.1 | - | 9057031 | ||||
BX479288.1 | - | |||||
DA492909.1 | - | |||||
EF173858.1 | - | |||||
ENST00000395762.0 | - | |||||
X52425.1 | - |