Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FLT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_004119.2 | NP_004110.2 | ||||
NR_130706.1 | ||||||
XM_011535015.2 | XP_011533317.1 | |||||
XM_011535017.2 | XP_011533319.1 | |||||
XM_011535018.2 | XP_011533320.1 | |||||
XM_017020486.1 | XP_016875975.1 | |||||
XM_017020487.1 | XP_016875976.1 | |||||
XM_017020488.1 | XP_016875977.1 | |||||
XM_017020489.1 | XP_016875978.1 | |||||
GenBank mRNA | AW057705.1 | - | ||||
AW772610.1 | - | |||||
BC036028.1 | - | |||||
BC126350.1 | - | 8992066 | ||||
BC144039.1 | - | 9052559 | ||||
BC144040.1 | - | 9052560 | ||||
BI461248.1 | - | |||||
ENST00000241453.0 | - | |||||
L36162.1 | - | |||||
U02687.1 | - | |||||
Z26652.1 | - |