Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_005233.5 | NP_005224.2 | ||||
NM_182644.2 | NP_872585.1 | |||||
XM_005264715.2 | XP_005264772.1 | |||||
XM_005264716.2 | XP_005264773.1 | |||||
GenBank mRNA | AF213459.1 | - | ||||
AF213460.1 | - | |||||
AK024352.1 | - | |||||
AK291411.1 | - | |||||
BC026247.1 | - | |||||
BC063282.1 | - | 30347891 | ||||
ENST00000336596.0 | - | |||||
HM437236.1 | - | |||||
M83941.1 | - |