Introducing: Accelerator™ Drug Development, 360˚ CDMO and CRO solutions

Explore now

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Microbe & ABR Detection
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            See other NTRK3 EBGEX Assays ›
            Gene Symbol:
            NTRK3
            Gene Name:
            neurotrophic receptor tyrosine kinase 3
            Gene Aliases:
            GP145-TrkC, TRKC, gp145(trkC)
            Chromosome Location:
            Chr.15: 87859749 - 88256796 on Build GRCh38
            UniGene ID:
            Hs.185701
            Species:
            Human
            Species Specific ID (Flybase ID):
            -
            Type:
            10
            Detection Label:
            Alexa Fluor 568
            ProductType:
            PrimeFlow Probe Set
            Assay ID VA10-3253344-PF
            Size 100 tests
            Availability Made To Order
            Catalog # PF-210
            Price (USD) 666.00
            Your Price
            Online offer:
            666.0
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data.
            ×
            Back To Top

            Assay Details

            Gene Symbol: NTRK3
            Interrogated Sequence Translated Protein IMAGE Clone ID
            RefSeq NM_001007156.2 NP_001007157.1
            NM_001012338.2 NP_001012338.1
            NM_001243101.1 NP_001230030.1
            NM_001320134.1 NP_001307063.1
            NM_001320135.1 NP_001307064.1
            NM_002530.3 NP_002521.2
            XM_006720543.3 XP_006720606.1
            XM_006720544.3 XP_006720607.1
            XM_006720545.3 XP_006720608.1
            XM_006720548.3 XP_006720611.1
            XM_006720549.3 XP_006720612.1
            XM_006720550.3 XP_006720613.1
            XM_011521637.2 XP_011519939.1
            XM_011521638.2 XP_011519940.1
            XM_017022240.1 XP_016877729.1
            XM_017022241.1 XP_016877730.1
            XM_017022242.1 XP_016877731.1
            XM_017022243.1 XP_016877732.1
            XM_017022244.1 XP_016877733.1
            XM_017022245.1 XP_016877734.1
            XM_017022246.1 XP_016877735.1
            XM_017022247.1 XP_016877736.1
            XM_017022248.1 XP_016877737.1
            XM_017022249.1 XP_016877738.1
            XM_017022250.1 XP_016877739.1
            XM_017022251.1 XP_016877740.1
            XM_017022252.1 XP_016877741.1
            XM_017022253.1 XP_016877742.1
            XM_017022254.1 XP_016877743.1
            GenBank mRNA AF052184.1 -
            AF058389.1 -
            AF058390.1 -
            AF125808.1 -
            AI613045.1 -
            AK094929.1 -
            AK297160.1 -
            AK302504.1 -
            AK309695.1 -
            AK313646.1 -
            AL109700.1 -
            AL134171.1 -
            AY065844.1 -
            BC013693.1 - 4181574
            BC128249.1 -
            BG741347.1 -
            BI962878.1 -
            BM675026.1 -
            BM887782.1 -
            BQ893817.1 -
            BT007291.1 -
            CB053605.1 -
            CD370531.1 -
            CK903553.1 -
            DA789502.1 -
            DQ323597.1 -
            ENST00000394480.0 -
            HY003065.1 -
            KJ534904.1 -
            KJ534905.1 -
            KJ534906.1 -
            KJ535052.1 -
            S76475.1 -
            S76476.1 -
            U05012.1 -

            Back To Top

            More Information


            Panther Classification:

            Molecular Function -

            transmembrane signal receptor

            Gene Ontology Categories:

            Function(s) Process(es)

            activation of MAPK activity
            neuron migration
            negative regulation of protein phosphorylation
            positive regulation of protein phosphorylation
            transmembrane receptor protein tyrosine kinase signaling pathway
            heart development
            circadian rhythm
            positive regulation of cell proliferation
            positive regulation of gene expression
            peptidyl-tyrosine phosphorylation
            modulation by virus of host transcription
            positive regulation of cell migration
            activation of protein kinase B activity
            positive regulation of peptidyl-serine phosphorylation
            neurotrophin signaling pathway
            mechanoreceptor differentiation
            positive regulation of apoptotic process
            response to ethanol
            protein autophosphorylation
            ephrin receptor signaling pathway
            neuron fate specification
            response to axon injury
            positive regulation of axon extension involved in regeneration
            negative regulation of astrocyte differentiation
            positive regulation of positive chemotaxis
            response to corticosterone
            positive regulation of synapse assembly
            negative regulation of cell death
            lens fiber cell differentiation
            cellular response to retinoic acid
            cochlea development
            activation of GTPase activity
            positive regulation of actin cytoskeleton reorganization
            p53 binding
            GPI-linked ephrin receptor activity
            neurotrophin receptor activity
            protein binding
            ATP binding
            neurotrophin binding

            Back To Top
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-6ff95d844f-mk26n:80/100.66.73.108:80.
            git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.28.0-Offline