Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_002015.3 | NP_002006.2 | ||||
XM_011535008.2 | XP_011533310.1 | |||||
XM_011535010.2 | XP_011533312.1 | |||||
GenBank mRNA | AF032885.1 | - | ||||
AK310511.1 | - | |||||
BC021981.2 | - | 2959021 | ||||
BC070065.1 | - | 30345006 | ||||
BT007455.1 | - | |||||
BX648278.1 | - | |||||
ENST00000379561.0 | - | |||||
U02310.1 | - | |||||
U36922.1 | - |