Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001304536.1 | NP_001291465.1 | ||||
NM_001304537.1 | NP_001291466.1 | |||||
NM_004438.4 | NP_004429.1 | |||||
XM_005246374.2 | XP_005246431.1 | |||||
GenBank mRNA | AA987900.1 | - | ||||
AF075040.1 | - | |||||
AK225181.1 | - | |||||
AK290306.1 | - | |||||
AK300772.1 | - | |||||
AK312380.1 | - | |||||
AL603465.1 | - | |||||
BC016981.1 | - | |||||
BC026327.1 | - | 4814429 | ||||
BC105000.1 | - | 8144003 | ||||
BC105002.1 | - | 8144005 | ||||
BM676153.1 | - | |||||
BM679073.1 | - | |||||
BM680394.1 | - | |||||
BM712812.1 | - | |||||
BM724253.1 | - | |||||
BQ009361.1 | - | |||||
DA451606.1 | - | |||||
ENST00000281821.0 | - | |||||
L36645.1 | - |