Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFATC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001278669.1 | NP_001265598.1 | ||||
NM_001278670.1 | NP_001265599.1 | |||||
NM_001278672.1 | NP_001265601.1 | |||||
NM_001278673.1 | NP_001265602.1 | |||||
NM_001278675.1 | NP_001265604.1 | |||||
NM_006162.4 | NP_006153.2 | |||||
NM_172387.2 | NP_765975.1 | |||||
NM_172388.2 | NP_765976.1 | |||||
NM_172389.2 | NP_765977.1 | |||||
NM_172390.2 | NP_765978.1 | |||||
XM_017025783.1 | XP_016881272.1 | |||||
GenBank mRNA | AA621079.1 | - | ||||
AA679278.1 | - | |||||
AK292641.1 | - | |||||
AK293760.1 | - | |||||
AK309546.1 | - | |||||
AW014910.1 | - | |||||
BC033848.1 | - | |||||
BC045628.1 | - | |||||
BC104753.1 | - | 8143756 | ||||
BC112243.1 | - | 8327711 | ||||
BC143457.1 | - | |||||
BF515172.1 | - | |||||
ENST00000329101.0 | - | |||||
EU887559.1 | - | |||||
EU887560.1 | - | |||||
EU887561.1 | - | |||||
EU887562.1 | - | |||||
EU887563.1 | - | |||||
EU887564.1 | - | |||||
EU887565.1 | - | |||||
EU887566.1 | - | |||||
HY039825.1 | - | |||||
U08015.1 | - | |||||
U59736.1 | - | |||||
U80917.1 | - | |||||
U80918.1 | - | |||||
U80919.1 | - | |||||
X99829.1 | - |