Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001303618.1 | NP_001290547.1 | ||||
NM_001303619.1 | NP_001290548.1 | |||||
NM_006566.3 | NP_006557.2 | |||||
XM_005266642.3 | XP_005266699.1 | |||||
XM_005266643.3 | XP_005266700.1 | |||||
XM_006722374.3 | XP_006722437.1 | |||||
XM_017025524.1 | XP_016881013.1 | |||||
XM_017025525.1 | XP_016881014.1 | |||||
XM_017025526.1 | XP_016881015.1 | |||||
XM_017025527.1 | XP_016881016.1 | |||||
GenBank mRNA | AJ537553.1 | - | ||||
AK313199.1 | - | |||||
BC074787.2 | - | 30915562 | ||||
DB224741.1 | - | |||||
ENST00000280200.0 | - | |||||
GU935335.1 | - | |||||
GU935336.1 | - | |||||
U56102.1 | - |