Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_002187.2 | NP_002178.2 | ||||
GenBank mRNA | AF180563.1 | - | ||||
AY046592.1 | - | |||||
AY046593.1 | - | |||||
BC067498.1 | - | 6971868 | ||||
BC067499.1 | - | 6971869 | ||||
BC067500.1 | - | 6971872 | ||||
BC067501.1 | - | 6971873 | ||||
BC067502.1 | - | 6971874 | ||||
BC074723.2 | - | 30915173 | ||||
EF173865.1 | - | |||||
ENST00000231228.0 | - | |||||
M65272.1 | - | |||||
M65290.1 | - |