Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000208.3 | NP_000199.2 | ||||
NM_001079817.2 | NP_001073285.1 | |||||
XM_011527988.2 | XP_011526290.2 | |||||
XM_011527989.2 | XP_011526291.2 | |||||
GenBank mRNA | AB208861.1 | - | ||||
AK025527.1 | - | |||||
AK300332.1 | - | |||||
AL365454.1 | - | |||||
BC047591.1 | - | |||||
BC117172.1 | - | 40125723 | ||||
BU684722.1 | - | |||||
ENST00000302850.0 | - | |||||
HY015976.1 | - | |||||
M10051.1 | - | |||||
M24555.1 | - | |||||
S70454.1 | - | |||||
X02160.1 | - |