Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001007565.2 | NP_001007566.1 | ||||
NM_001195478.1 | NP_001182407.1 | |||||
NM_001195479.1 | NP_001182408.1 | |||||
NM_006070.5 | NP_006061.2 | |||||
XM_005247066.1 | XP_005247123.1 | |||||
XM_006713472.1 | XP_006713535.1 | |||||
XM_006713473.1 | XP_006713536.1 | |||||
XM_011512334.1 | XP_011510636.1 | |||||
XM_017005527.1 | XP_016861016.1 | |||||
XM_017005528.1 | XP_016861017.1 | |||||
XM_017005529.1 | XP_016861018.1 | |||||
XM_017005530.1 | XP_016861019.1 | |||||
GenBank mRNA | AB097040.1 | - | ||||
AB097041.1 | - | |||||
AB731569.1 | - | |||||
AB731570.1 | - | |||||
AK093456.1 | - | |||||
AK123588.1 | - | |||||
AK127920.1 | - | |||||
BC001483.2 | - | 3507440 | ||||
BC009241.2 | - | 2962943 | ||||
BC023599.2 | - | 4555507 | ||||
BC041600.1 | - | 5458899 | ||||
BE409447.1 | - | |||||
BM746125.1 | - | |||||
BQ008360.1 | - | |||||
BT007428.1 | - | |||||
CR456781.1 | - | |||||
DA704111.1 | - | |||||
Y07968.1 | - |