Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001128215.1 | NP_001121687.1 | ||||
XM_011539748.2 | XP_011538050.1 | |||||
XM_011539751.2 | XP_011538053.1 | |||||
XM_011539752.2 | XP_011538054.1 | |||||
XM_011539753.2 | XP_011538055.1 | |||||
GenBank mRNA | BC157888.1 | - | 8862709 | |||
BC171908.1 | - | 9054562 | ||||
EF426484.1 | - | |||||
ENST00000404743.0 | - |