Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAIP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_020459.1 | NP_065192.1 | ||||
XM_005264310.3 | XP_005264367.1 | |||||
XM_005264311.4 | XP_005264368.1 | |||||
XM_011532842.2 | XP_011531144.1 | |||||
XM_011532843.2 | XP_011531145.1 | |||||
GenBank mRNA | AB032981.1 | - | ||||
AL110211.1 | - | |||||
DA247526.1 | - | |||||
DA346299.1 | - | |||||
ENST00000244221.0 | - |