Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctla4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001281976.1 | NP_001268905.1 | ||||
NM_009843.4 | NP_033973.2 | |||||
GenBank mRNA | AB097214.1 | - | ||||
AI323635.1 | - | |||||
AK138891.1 | - | |||||
BC042741.1 | - | 4936052 | ||||
BC052683.1 | - | 30072821 | ||||
U90270.1 | - | |||||
X05719.1 | - |