Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_011577.1 | NP_035707.1 | ||||
GenBank mRNA | AJ009862.1 | - | ||||
AK144163.1 | - | |||||
BC013738.1 | - | 3586216 | ||||
M13177.1 | - |