Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_010229.2 | NP_034359.2 | ||||
GenBank mRNA | AK045865.1 | - | ||||
AK149292.1 | - | |||||
AK163640.1 | - | |||||
BC109003.1 | - | |||||
BC109004.1 | - | |||||
L36163.1 | - | |||||
M64689.1 | - | |||||
X59398.1 | - |