Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_010140.3 | NP_034270.1 | ||||
GenBank mRNA | AK041935.1 | - | ||||
AK044936.1 | - | |||||
AK045219.1 | - | |||||
AK084921.1 | - | |||||
AK089119.1 | - | |||||
AK132335.1 | - | |||||
BC053479.1 | - | |||||
BC060164.1 | - | |||||
BC093483.1 | - | |||||
BC138641.1 | - | 8861661 | ||||
BC138643.1 | - | 8861663 | ||||
BC145349.1 | - | 9053905 | ||||
M68513.1 | - | |||||
M68515.1 | - |