Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001166361.1 | NP_001159833.1 | ||||
NM_001166362.1 | NP_001159834.1 | |||||
NM_001166363.1 | NP_001159835.1 | |||||
NM_010205.2 | NP_034335.1 | |||||
GenBank mRNA | AK131980.1 | - | ||||
BC048734.1 | - | 6513131 | ||||
BU512456.1 | - | |||||
D12482.1 | - | |||||
D12483.1 | - | |||||
HQ260698.1 | - | |||||
U18673.1 | - | |||||
Z48746.1 | - |