Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001290424.1 | NP_001277353.1 | ||||
NM_008031.3 | NP_032057.2 | |||||
GenBank mRNA | AF170530.1 | - | ||||
AF179463.1 | - | |||||
AF461114.1 | - | |||||
AK046605.1 | - | |||||
AK053211.1 | - | |||||
AK053701.1 | - | |||||
AK053829.1 | - | |||||
AK080898.1 | - | |||||
AK080948.1 | - | |||||
AK140343.1 | - | |||||
AK208300.1 | - | |||||
BC079671.1 | - | 30532682 | ||||
L23971.1 | - |