Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nup210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_018815.2 | NP_061285.2 | ||||
GenBank mRNA | AF113751.1 | - | ||||
AK036378.1 | - | |||||
AK053417.1 | - | |||||
AK077812.1 | - | |||||
AK078913.1 | - | |||||
AK090317.1 | - | |||||
AK157272.1 | - | |||||
AK173056.1 | - | |||||
AK202156.1 | - | |||||
BC020091.1 | - | |||||
BC052468.1 | - | 6835038 | ||||
DT908828.1 | - |