Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
GenBank mRNA | AF128214.1 | - | ||||
AF128215.1 | - | |||||
AK162981.1 | - | |||||
BC103608.1 | - | 40046200 | ||||
BC103609.1 | - | 40046202 | ||||
BC103610.1 | - | 40046204 | ||||
BC103614.1 | - | 40046206 | ||||
BY154173.1 | - | |||||
M86671.1 | - |