Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_019739.3 | NP_062713.2 | ||||
GenBank mRNA | AF114258.1 | - | ||||
AF126056.1 | - | |||||
AJ252157.1 | - | |||||
AK029013.1 | - | |||||
AK030304.1 | - | |||||
AK039265.1 | - | |||||
AK043583.1 | - | |||||
AK053633.1 | - | |||||
AK054289.1 | - | |||||
AK137629.1 | - | |||||
AK154041.1 | - | |||||
AK185432.1 | - | |||||
AK192846.1 | - | |||||
AK211466.1 | - | |||||
AK211487.1 | - | |||||
AK212278.1 | - | |||||
AK212379.1 | - |