Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_007936.3 | NP_031962.2 | ||||
GenBank mRNA | AK013481.1 | - | ||||
AK028509.1 | - | |||||
AK132203.1 | - | |||||
AK147698.1 | - | |||||
BC004782.1 | - | 3588285 | ||||
BC052164.1 | - | 6512978 | ||||
S57168.1 | - | |||||
X57241.1 | - | |||||
X65138.1 | - |